COMH042350EA Combi-Blocks, Inc.

4-Chloro-1-ethyl-5-methyl-1h-pyrazole

  • MDL number MFCD12026907
  • Cas Number : 1172915-66-0
  • Molecular Weight : 144.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0571
Availability details for
Quantity
  • Availability
  • Add to Cart