COMH042351B2 Combi-Blocks, Inc.

N-(3-Methylphenyl)-n-(phenylsulfonyl)glycine

  • MDL number MFCD02576171
  • Cas Number : 117309-37-2
  • Molecular Weight : 305.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0771

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0771
Availability details for
Quantity
  • Availability
  • Add to Cart