COMH04235232 Combi-Blocks, Inc.

[3-(1-Isopropyl-1h-benzimidazol-2-yl)propyl]amine dihydrochloride

  • MDL number MFCD17430444
  • Cas Number : 1245570-14-2
  • Molecular Weight : 290.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0899
Availability details for
Quantity
  • Availability
  • Add to Cart