COMH04235234 Combi-Blocks, Inc.

2-((2-[(4-Chloro-2-nitrophenyl)amino]ethyl)amino)ethanol

  • MDL number MFCD00640770
  • Cas Number : 282104-85-2
  • Molecular Weight : 259.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-0901
Availability details for
Quantity
  • Availability
  • Add to Cart