COMH042352A5 Combi-Blocks, Inc.

Ethyl 2-isothiocyanato-5,6,7,8-tetrahydro-4h-cyclohepta[b]thiophene-3-carboxylate

  • MDL number MFCD03945301
  • Cas Number : 257610-91-6
  • Molecular Weight : 281.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1392

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1392
Availability details for
Quantity
  • Availability
  • Add to Cart