COMH042352D8 Combi-Blocks, Inc.

1-[2-(1,3-Dioxo-1,3-dihydro-2h-isoindol-2-yl)ethyl]-1h-indole-2,3-dione

  • MDL number MFCD00469808
  • Cas Number : 303065-17-0
  • Molecular Weight : 320.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1601
Availability details for
Quantity
  • Availability
  • Add to Cart