COMH0423535E Combi-Blocks, Inc.

4-Amino-5-(5-bromo-2-fluorophenyl)-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD10691190
  • Cas Number : 1152520-73-4
  • Molecular Weight : 289.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2474
Availability details for
Quantity
  • Availability
  • Add to Cart