COMH04235365 Combi-Blocks, Inc.

N-(4-Chlorophenyl)-n-(methylsulfonyl)alanine

  • MDL number MFCD05815151
  • Cas Number : 1008074-24-5
  • Molecular Weight : 277.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2481
Availability details for
Quantity
  • Availability
  • Add to Cart