COMH04235377 Combi-Blocks, Inc.

2-(5-Chloro-2-thienyl)-7,8-dimethylquinoline-4-carbonyl chloride

  • MDL number MFCD12198069
  • Cas Number : 1160261-41-5
  • Molecular Weight : 336.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2499
Availability details for
Quantity
  • Availability
  • Add to Cart