COMH042353CF Combi-Blocks, Inc.

2'-Sec-butyl-1'-oxo-1',4'-dihydro-2'h-spiro[cyclohexane-1,3'-isoquinoline]-4'-carboxylic acid

  • MDL number MFCD18446996
  • Cas Number : 1269525-47-4
  • Molecular Weight : 315.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2587
Availability details for
Quantity
  • Availability
  • Add to Cart