COMH0423543E Combi-Blocks, Inc.

tert-Butyl 8-bromo-5-methyl-1,3,4,5-tetrahydro-2h-pyrido[4,3-b]indole-2-carboxylate

  • MDL number MFCD29034971
  • Cas Number : 1173155-41-3
  • Molecular Weight : 365.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2698
Availability details for
Quantity
  • Availability
  • Add to Cart