COMH04235447 Combi-Blocks, Inc.

5-(Methoxymethyl)-1h-1,2,4-triazole

  • MDL number MFCD20502781
  • Cas Number : 23159-59-3
  • Molecular Weight : 113.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2707
Availability details for
Quantity
  • Availability
  • Add to Cart