COMH04235455 Combi-Blocks, Inc.

[2-(4-Chloro-2-isopropyl-5-methylphenoxy)ethyl]amine hydrochloride

  • MDL number MFCD08072593
  • Cas Number : 1051368-66-1
  • Molecular Weight : 264.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2721
Availability details for
Quantity
  • Availability
  • Add to Cart