COMH042354A1 Combi-Blocks, Inc.

Methyl n-(3,4-dimethylphenyl)-n-(methylsulfonyl)alaninate

  • MDL number MFCD05811692
  • Cas Number : 1008051-01-1
  • Molecular Weight : 285.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2797
Availability details for
Quantity
  • Availability
  • Add to Cart