COMH042354B7 Combi-Blocks, Inc.

3-[(4-Chlorobenzyl)oxy]benzoyl chloride

  • MDL number MFCD12197999
  • Cas Number : 1160260-07-0
  • Molecular Weight : 281.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2819
Availability details for
Quantity
  • Availability
  • Add to Cart