COMH0423550A Combi-Blocks, Inc.

1-((2-[(4-Chloro-2-nitrophenyl)amino]ethyl)amino)propan-2-ol

  • MDL number MFCD01203160
  • Cas Number : 330634-22-5
  • Molecular Weight : 273.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2902

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2902
Availability details for
Quantity
  • Availability
  • Add to Cart