COMH0423551B Combi-Blocks, Inc.

3-(3,4-Dihydroisoquinolin-2(1h)-yl)butanoic acid

  • MDL number MFCD12132203
  • Cas Number : 1153828-35-3
  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2919
Availability details for
Quantity
  • Availability
  • Add to Cart