COMH04235530 Combi-Blocks, Inc.

3-[(Dimethylamino)carbonothioyl]aminobenzoic acid

  • MDL number MFCD29762483
  • Cas Number : 1273305-86-4
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2940

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2940
Availability details for
Quantity
  • Availability
  • Add to Cart