COMH0423553C Combi-Blocks, Inc.

Ethyl 2-(4-methoxybenzyl)piperidine-2-carboxylate hydrochloride

  • MDL number MFCD28505154
  • Cas Number : 1993278-30-0
  • Molecular Weight : 313.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2952
Availability details for
Quantity
  • Availability
  • Add to Cart