COMH0423554B Combi-Blocks, Inc.

6-Chloro-2-(2-thienyl)quinoline-4-carbonyl chloride

  • MDL number MFCD03421277
  • Cas Number : 1160263-06-8
  • Molecular Weight : 308.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2967
Availability details for
Quantity
  • Availability
  • Add to Cart