COMH0423554F Combi-Blocks, Inc.

N-Ethyl-3,4-dimethylaniline

  • MDL number MFCD03210677
  • Cas Number : 27285-20-7
  • Molecular Weight : 149.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2971
Availability details for
Quantity
  • Availability
  • Add to Cart