COMH04235557 Combi-Blocks, Inc.

N-(1,3-Thiazol-5-ylmethyl)propan-2-amine dihydrochloride

  • MDL number MFCD18071380
  • Cas Number : 1332530-31-0
  • Molecular Weight : 229.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2979

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2979
Availability details for
Quantity
  • Availability
  • Add to Cart