COMH04235568 Combi-Blocks, Inc.

4-(5-[(Cyclobutylcarbonyl)amino]pyridin-2-yl)benzoic acid

  • MDL number MFCD14281922
  • Cas Number : 1170357-94-4
  • Molecular Weight : 296.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2997

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2997
Availability details for
Quantity
  • Availability
  • Add to Cart