COMH0423556F Combi-Blocks, Inc.

Ethyl 3-amino-5-(3-methylphenyl)thiophene-2-carboxylate

  • MDL number MFCD09835651
  • Cas Number : 1019012-08-8
  • Molecular Weight : 261.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3004
Availability details for
Quantity
  • Availability
  • Add to Cart