COMH0423558C Combi-Blocks, Inc.

(2R,4R)-3-Acetyl-2-(4-fluorophenyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD13248706
  • Cas Number : 110078-80-3
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3033
Availability details for
Quantity
  • Availability
  • Add to Cart