COMH042355E6 Combi-Blocks, Inc.

[1-Cyclopropyl-2-(2,4-dimethylphenoxy)ethyl]amine

  • MDL number MFCD19605870
  • Cas Number : 1340391-41-4
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3123
Availability details for
Quantity
  • Availability
  • Add to Cart