COMH042355F4 Combi-Blocks, Inc.

N-(1H-Indol-5-ylmethyl)-n-(pyridin-3-ylmethyl)amine dihydrochloride

  • MDL number MFCD12026960
  • Cas Number : 1185295-06-0
  • Molecular Weight : 310.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3137

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3137
Availability details for
Quantity
  • Availability
  • Add to Cart