COMH0423562B Combi-Blocks, Inc.

1-Anilino-2-methylpropan-2-ol

  • MDL number MFCD11934537
  • Cas Number : 127832-84-2
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3192

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3192
Availability details for
Quantity
  • Availability
  • Add to Cart