COMH042356A1 Combi-Blocks, Inc.

6-Chloro-n,n'-bis(3-chloro-4-methylphenyl)-1,3,5-triazine-2,4-diamine

  • MDL number MFCD22374951
  • Cas Number : 1379811-60-5
  • Molecular Weight : 394.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3310
Availability details for
Quantity
  • Availability
  • Add to Cart