COMH042356B7 Combi-Blocks, Inc.

4-(3-(2-[(tert-Butoxycarbonyl)(methyl)amino]ethyl)-1,2,4-oxadiazol-5-yl)benzoic acid

  • MDL number MFCD12028376
  • Cas Number : 1142210-76-1
  • Molecular Weight : 347.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3332

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3332
Availability details for
Quantity
  • Availability
  • Add to Cart