COMH042356BC Combi-Blocks, Inc.

5-(4-Butoxyphenyl)-1-(4-ethylphenyl)-3-[(4-ethylphenyl)amino]-1,5-dihydro-2h-pyrrol-2-one

  • MDL number MFCD01439999
  • Cas Number : 312517-96-7
  • Molecular Weight : 454.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3337
Availability details for
Quantity
  • Availability
  • Add to Cart