COMH042356C0 Combi-Blocks, Inc.

5-Chloro-1,3-dimethyl-6-propyl-1,6-dihydro-7h-pyrazolo[4,3-d]pyrimidin-7-one

  • MDL number MFCD22056540
  • Cas Number : 1365964-11-9
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3341
Availability details for
Quantity
  • Availability
  • Add to Cart