COMH042356CA Combi-Blocks, Inc.

2-(1,1'-Biphenyl-4-yloxy)-2-methylpropanoic acid

  • MDL number MFCD03422292
  • Cas Number : 1222-74-8
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3351
Availability details for
Quantity
  • Availability
  • Add to Cart