COMH04235744 Combi-Blocks, Inc.

3-(Cyclohexylamino)pyrrolidine-2,5-dione

  • MDL number MFCD12099304
  • Cas Number : 1189013-16-8
  • Molecular Weight : 196.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3473
Availability details for
Quantity
  • Availability
  • Add to Cart