COMH04235768 Combi-Blocks, Inc.

1-[7-[(E)-2-(Dimethylamino)vinyl]-2-(ethylthio)[1,2,4]triazolo[1,5-a]pyrimidin-6-yl]ethanone

  • MDL number MFCD19103621
  • Cas Number : 1306753-63-8
  • Molecular Weight : 291.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3509
Availability details for
Quantity
  • Availability
  • Add to Cart