COMH04235788 Combi-Blocks, Inc.

2-(3-Methyl-1h-pyrazol-1-yl)propanohydrazide

  • MDL number MFCD03419640
  • Cas Number : 1005695-50-0
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3542
Availability details for
Quantity
  • Availability
  • Add to Cart