COMH042357A0 Combi-Blocks, Inc.

7-Chloro-2-(3,4-dimethylphenyl)-8-methylquinoline-4-carbonyl chloride

  • MDL number MFCD03422872
  • Cas Number : 1160256-40-5
  • Molecular Weight : 344.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3566
Availability details for
Quantity
  • Availability
  • Add to Cart