COMH042357B5 Combi-Blocks, Inc.

Diethyl 2-aminoterephthalate

  • MDL number MFCD00272347
  • Cas Number : 27210-70-4
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3587
Availability details for
Quantity
  • Availability
  • Add to Cart