COMH042357E4 Combi-Blocks, Inc.

5-(2-Hydroxyphenyl)isoxazole-3-carboxylic acid

  • MDL number MFCD05237206
  • Cas Number : 33282-12-1
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3634
Availability details for
Quantity
  • Availability
  • Add to Cart