COMH04235875 Combi-Blocks, Inc.

Methyl 3-(1,1-dioxido-3-oxo-1,2-benzisothiazol-2(3h)-yl)propanoate

  • MDL number MFCD10660120
  • Cas Number : 22010-99-7
  • Molecular Weight : 269.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3779
Availability details for
Quantity
  • Availability
  • Add to Cart