COMH042358C0 Combi-Blocks, Inc.

4-(2-Cyano-acetylamino)-benzoic acid ethyl ester

  • MDL number MFCD00488476
  • Cas Number : 15029-53-5
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3854
Availability details for
Quantity
  • Availability
  • Add to Cart