COMH042358D9 Combi-Blocks, Inc.

2,2-Dimethyl-3-[(1,3-thiazol-2-ylamino)carbonyl]cyclopropanecarboxylic acid

  • MDL number MFCD20425963
  • Cas Number : 1344039-20-8
  • Molecular Weight : 240.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3879
Availability details for
Quantity
  • Availability
  • Add to Cart