COMH042358E3 Combi-Blocks, Inc.

3-(5-([(4-Methoxyphenyl)amino]carbonyl)-1,3,4-thiadiazol-2-yl)propanoic acid

  • MDL number MFCD12028267
  • Cas Number : 1142209-61-7
  • Molecular Weight : 307.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3889
Availability details for
Quantity
  • Availability
  • Add to Cart