COMH04235929 Combi-Blocks, Inc.

2-(4-Formylphenoxy)acetamide

  • MDL number MFCD01050440
  • Cas Number : 135857-20-4
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3959
Availability details for
Quantity
  • Availability
  • Add to Cart