COMH0423593C Combi-Blocks, Inc.

1-[(3-Bromophenoxy)methyl]-1h-pyrazole-3-carboxylic acid

  • MDL number MFCD04967929
  • Cas Number : 1006487-34-8
  • Molecular Weight : 297.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3978

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-3978
Availability details for
Quantity
  • Availability
  • Add to Cart