COMH04235960 Combi-Blocks, Inc.

1-(1,3-Benzothiazol-2-yl)pentane-2,4-dione

  • MDL number MFCD22374953
  • Cas Number : 1379811-76-3
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4014
Availability details for
Quantity
  • Availability
  • Add to Cart