COMH042359C5 Combi-Blocks, Inc.

Dimethyl 2-(2,2,6-trimethyl-3-thioxo-2,3-dihydroquinolin-4(1h)-ylidene)-1,3-dithiole-4,5-dicarboxylate

  • MDL number MFCD00624654
  • Cas Number : 258267-12-8
  • Molecular Weight : 421.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4115
Availability details for
Quantity
  • Availability
  • Add to Cart