COMH042359C9 Combi-Blocks, Inc.

1-Allyl-5-chloro-1h-1,2,4-triazole

  • MDL number MFCD27977470
  • Cas Number : 1573547-66-6
  • Molecular Weight : 143.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4119
Availability details for
Quantity
  • Availability
  • Add to Cart