COMH04235A0A Combi-Blocks, Inc.

(4-[(3-Nitropyridin-2-yl)oxy]phenyl)amine

  • MDL number MFCD06825490
  • Cas Number : 102766-76-7
  • Molecular Weight : 231.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4184

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4184
Availability details for
Quantity
  • Availability
  • Add to Cart