COMH04235A27 Combi-Blocks, Inc.

3-Bromo-1-(4-fluorophenyl)-1h-pyrrole-2,5-dione

  • MDL number MFCD01935482
  • Cas Number : 279686-72-5
  • Molecular Weight : 270.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4213

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4213
Availability details for
Quantity
  • Availability
  • Add to Cart